ENA432539711 Enamine

2-cyclopentylidenepropanedinitrile

  • MDL number MFCD00774818
  • Cas Number : 5660-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195207
Availability details for
Quantity
  • Availability
  • Add to Cart