ENA432539712 Enamine

3-[(2-hydroxyphenyl)formamido]propanoic acid

  • MDL number MFCD09047510
  • Cas Number : 6292-94-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196656

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196656
Availability details for
Quantity
  • Availability
  • Add to Cart