ENA432539714 Enamine

5-bromo-2-nitropyridine-4-carboxylic acid

  • MDL number MFCD21648462
  • Cas Number : 1356412-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196680

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196680
Availability details for
Quantity
  • Availability
  • Add to Cart