ENA432539717 Enamine

2-iodo-4-(methylsulfanyl)benzonitrile

  • MDL number MFCD22489435
  • Cas Number : 54558-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195208
Availability details for
Quantity
  • Availability
  • Add to Cart