ENA432539721 Enamine

5-amino-1-(2,2-diethoxyethyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD28145377
  • Cas Number : 135829-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196688

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196688
Availability details for
Quantity
  • Availability
  • Add to Cart