ENA432539724 Enamine

3-amino-2-cyanobut-2-enethioamide

  • Cas Number : 758721-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196692
Availability details for
Quantity
  • Availability
  • Add to Cart