ENA432539725 Enamine

3-(3-chlorophenyl)-2-formamidopropanoic acid

  • MDL number MFCD19661103
  • Cas Number : 1341669-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196694

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196694
Availability details for
Quantity
  • Availability
  • Add to Cart