ENA432539727 Enamine

2,2-dimethyl-3,4-dihydro-2H-1-benzopyran-4,5-diol

  • MDL number MFCD24714852
  • Cas Number : 100942-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196716

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196716
Availability details for
Quantity
  • Availability
  • Add to Cart