ENA432539730 Enamine

3-fluoro-2-(methylsulfanyl)benzoic acid

  • MDL number MFCD16836264
  • Cas Number : 147460-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196723

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196723
Availability details for
Quantity
  • Availability
  • Add to Cart