ENA432539734 Enamine

2-[4-(trifluoromethyl)phenyl]ethane-1-sulfonyl chloride

  • MDL number MFCD19372884
  • Cas Number : 721-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196734

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196734
Availability details for
Quantity
  • Availability
  • Add to Cart