ENA432539736 Enamine

2-methanesulfonyl-1-(4-methylphenyl)propan-1-amine hydrochloride

  • MDL number MFCD28145380
  • Cas Number : 1803591-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196736

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196736
Availability details for
Quantity
  • Availability
  • Add to Cart