ENA432539745 Enamine

4-trifluoromethanesulfinylbenzoic acid

  • MDL number MFCD12773913
  • Cas Number : 948-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196783

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196783
Availability details for
Quantity
  • Availability
  • Add to Cart