ENA432539747 Enamine

5-fluoro-1-methyl-1H-indole-3-carboxylic acid

  • MDL number MFCD15523396
  • Cas Number : 310886-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218-222
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195216
Availability details for
Quantity
  • Availability
  • Add to Cart