ENA432539751 Enamine

2-amino-N-[2-(2-methoxyphenyl)ethyl]acetamide hydrochloride

  • MDL number MFCD28145386
  • Cas Number : 1803592-00-8
  • Molecular Weight : 244.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-N-(2-methoxyphenethyl)acetamide hydrochloride
Inchi Code 1S/C11H16N2O2.ClH/c1-15-10-5-3-2-4-9(10)6-7-13-11(14)8-12;/h2-5H,6-8,12H2,1H3,(H,13,14);1H
Inchi Key KGENCZVNJOZDTC-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-196789

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-196789
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart