ENA432539754 Enamine

(2-aminoethyl)(pentan-3-yl)amine

  • MDL number MFCD04360041
  • Cas Number : 129394-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195219
Availability details for
Quantity
  • Availability
  • Add to Cart