ENA432539757 Enamine

1-[4-bromo-3-(trifluoromethyl)phenyl]cyclopropan-1-ol

  • MDL number MFCD24148366
  • Cas Number : 1539874-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196802

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196802
Availability details for
Quantity
  • Availability
  • Add to Cart