ENA432539760 Enamine

5-ethynylpyridine-3-carbonitrile

  • MDL number MFCD11109673
  • Cas Number : 152803-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194908

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194908
Availability details for
Quantity
  • Availability
  • Add to Cart