ENA432539761 Enamine

[2-methoxy-6-(trifluoromethyl)pyridin-3-yl]boronic acid

  • MDL number MFCD10696221
  • Cas Number : 1429874-11-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196822
Availability details for
Quantity
  • Availability
  • Add to Cart