ENA432539762 Enamine

2-{[(tert-butoxy)carbonyl]amino}-5,5,5-trifluoropentanoic acid

  • MDL number MFCD18910160
  • Cas Number : 1454913-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195222

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195222
Availability details for
Quantity
  • Availability
  • Add to Cart