ENA432539768 Enamine

N-methylprop-2-ene-1-sulfonamide

  • MDL number MFCD23742749
  • Cas Number : 104459-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196855

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196855
Availability details for
Quantity
  • Availability
  • Add to Cart