ENA432539769 Enamine

1,3-difluoro-5-(prop-2-yn-1-yloxy)benzene

  • MDL number MFCD17216189
  • Cas Number : 457628-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196856

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196856
Availability details for
Quantity
  • Availability
  • Add to Cart