ENA432539770 Enamine

1-bromo-3-(bromomethyl)-2,4-difluorobenzene

  • MDL number MFCD27950792
  • Cas Number : 438050-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196866

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196866
Availability details for
Quantity
  • Availability
  • Add to Cart