ENA432539771 Enamine

8-fluoro-2-methyl-4H-3,1-benzoxazin-4-one

  • MDL number MFCD20040002
  • Cas Number : 1044749-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196867
Availability details for
Quantity
  • Availability
  • Add to Cart