ENA432539774 Enamine

N-methyl-4-(sulfamoylmethyl)benzamide

  • MDL number MFCD16671907
  • Cas Number : 1247384-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195226
Availability details for
Quantity
  • Availability
  • Add to Cart