ENA432539789 Enamine

2-(3,5-dimethyl-1H-pyrazol-1-yl)-6-methylpyrimidin-4-ol

  • MDL number MFCD00097142
  • Cas Number : 65004-42-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-137
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196918

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196918
Availability details for
Quantity
  • Availability
  • Add to Cart