ENA432539800 Enamine

(3-methyl-1,2,4-thiadiazol-5-yl)methanol

  • MDL number MFCD22393840
  • Cas Number : 1803600-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-195247
Availability details for
Quantity
  • Availability
  • Add to Cart