ENA432539804 Enamine

4-chloro-2-fluoro-N-methylaniline hydrochloride

  • MDL number MFCD28145412
  • Cas Number : 956828-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196933

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196933
Availability details for
Quantity
  • Availability
  • Add to Cart