ENA432539810 Enamine

N,N,4-trimethylpyrimidin-2-amine

  • MDL number MFCD18816628
  • Cas Number : 99357-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196945

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196945
Availability details for
Quantity
  • Availability
  • Add to Cart