ENA432539817 Enamine

5-bromo-N,2-dimethyl-N-[(3-methylphenyl)methyl]thiophene-3-sulfonamide

  • MDL number MFCD28139607
  • Cas Number : 1507874-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195263

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195263
Availability details for
Quantity
  • Availability
  • Add to Cart