ENA432539832 Enamine

(5-chloro-4-iodo-2-methoxyphenyl)methanamine hydrochloride

  • MDL number MFCD28139610
  • Cas Number : 1803581-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195315

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195315
Availability details for
Quantity
  • Availability
  • Add to Cart