ENA432539836 Enamine

2-(cyclopropylmethoxy)pyridine-4-carbothioamide

  • MDL number MFCD11136716
  • Cas Number : 1019546-78-1
  • Molecular Weight : 208.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(cyclopropylmethoxy)-4-pyridinecarbothioamide
Inchi Code 1S/C10H12N2OS/c11-10(14)8-3-4-12-9(5-8)13-6-7-1-2-7/h3-5,7H,1-2,6H2,(H2,11,14)
Inchi Key WSGSPSNQKWDXEZ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-195317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195317
Availability details for
Quantity
  • Availability
  • Add to Cart