ENA432539837 Enamine

2,2-dimethyl-1,8-dioxa-4-azaspiro[5.5]undecane

  • MDL number MFCD21127115
  • Cas Number : 1495198-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195318

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195318
Availability details for
Quantity
  • Availability
  • Add to Cart