ENA432539842 Enamine

methyl N-(3-amino-2-methylphenyl)carbamate

  • MDL number MFCD11128936
  • Cas Number : 157563-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195323

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195323
Availability details for
Quantity
  • Availability
  • Add to Cart