ENA432539843 Enamine

N2-methyl-N2-[(4-methylphenyl)methyl]-1,3-benzoxazole-2,5-diamine

  • MDL number MFCD28139612
  • Cas Number : 1502768-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195324

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195324
Availability details for
Quantity
  • Availability
  • Add to Cart