ENA432539845 Enamine

5-phenylpyrimidin-2-amine

  • MDL number MFCD01646362
  • Cas Number : 31408-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159-161
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195327

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195327
Availability details for
Quantity
  • Availability
  • Add to Cart