ENA432539852 Enamine

ethyl 2-[(6-formyl-1,3-dioxaindan-5-yl)oxy]acetate

  • MDL number MFCD16283831
  • Cas Number : 13668-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195339

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195339
Availability details for
Quantity
  • Availability
  • Add to Cart