ENA432539853 Enamine

methyl 2-[2-(hydroxymethyl)-1,3-thiazol-4-yl]acetate

  • MDL number MFCD26846055
  • Cas Number : 1565348-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195341
Availability details for
Quantity
  • Availability
  • Add to Cart