ENA432539856 Enamine

2-(4-bromo-3-fluorophenyl)pyrrolidine

  • MDL number MFCD11848581
  • Cas Number : 1175528-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195354
Availability details for
Quantity
  • Availability
  • Add to Cart