ENA432539872 Enamine

2-(2-aminophenyl)-2-oxoethane-1-sulfonamide

  • MDL number MFCD24631979
  • Cas Number : 49666-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195374

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195374
Availability details for
Quantity
  • Availability
  • Add to Cart