ENA432539873 Enamine

2-(propan-2-yl)furan-3-carboxylic acid

  • MDL number MFCD20639748
  • Cas Number : 99698-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195375

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195375
Availability details for
Quantity
  • Availability
  • Add to Cart