ENA432539876 Enamine

methyl 1-acetyl-3,3-dimethoxycyclobutane-1-carboxylate

  • MDL number MFCD21950420
  • Cas Number : 1499341-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195386
Availability details for
Quantity
  • Availability
  • Add to Cart