ENA432539878 Enamine

4-(3-amino-4-bromo-1H-pyrazol-1-yl)-3-fluorobenzonitrile

  • MDL number MFCD14594856
  • Cas Number : 1247586-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195394

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195394
Availability details for
Quantity
  • Availability
  • Add to Cart