ENA432539885 Enamine

3-methyl-N-(3-methylbutan-2-yl)pyridin-4-amine

  • MDL number MFCD19675158
  • Cas Number : 1343678-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195406

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195406
Availability details for
Quantity
  • Availability
  • Add to Cart