ENA432539890 Enamine

(1-cyclohexylpyrrolidin-2-yl)methanamine

  • MDL number MFCD18629315
  • Cas Number : 54929-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197056

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197056
Availability details for
Quantity
  • Availability
  • Add to Cart