ENA432539893 Enamine

2-(3-aminocyclopentyl)-octahydropyrrolo[1,2-a]piperazin-6-one

  • MDL number MFCD24318703
  • Cas Number : 1543883-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197097
Availability details for
Quantity
  • Availability
  • Add to Cart