ENA432539899 Enamine

2-chloro-1-{4-chloro-1H-pyrrolo[2,3-b]pyridin-3-yl}ethan-1-one

  • MDL number MFCD24222233
  • Cas Number : 1553611-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197105

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197105
Availability details for
Quantity
  • Availability
  • Add to Cart