ENA432539908 Enamine

7-chloroquinolin-2-amine

  • MDL number MFCD11877922
  • Cas Number : 43200-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195433

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195433
Availability details for
Quantity
  • Availability
  • Add to Cart