ENA432539909 Enamine

N-(1,3-dioxaindan-5-yl)-2-hydroxypropanamide

  • MDL number MFCD18156711
  • Cas Number : 1241040-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195434

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195434
Availability details for
Quantity
  • Availability
  • Add to Cart