ENA432539925 Enamine

N2-[(2-chlorophenyl)methyl]-1,3-benzoxazole-2,6-diamine

  • MDL number MFCD22890153
  • Cas Number : 1518095-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195437

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195437
Availability details for
Quantity
  • Availability
  • Add to Cart