ENA432539927 Enamine

2-[(6-nitroquinolin-2-yl)amino]propanoic acid

  • MDL number MFCD12468669

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195438

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195438
Availability details for
Quantity
  • Availability
  • Add to Cart